• USA Home
  • SAB4700756 - Monoclonal Anti-CPNE7 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB4700756 Sigma

Monoclonal Anti-CPNE7 antibody produced in mouse

clone CPNE7-01, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-copine VII

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, CO-CP, Primary Antibodies
species reactivity   human
application(s)   immunoblotting: suitable
clone   CPNE7-01, monoclonal
concentration   1 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG
shipped in   wet ice
storage temp.   2-8°C
Gene Information   human ... CPNE7(27132)
biological source   mouse
conjugate   unconjugated
NCBI accession no.   NM_014427.4

Description

Immunogen

Bacterially produced GST-fused C terminus of human CPNE7.

General description

The antibody CPNE7-01 recognizes C terminus of human CPNE7, a calcium-binding cytoplasmic protein expressed mainly in brain.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?