• USA Home
  • SAB4700772 - Monoclonal Anti-Neurofilament medium protein antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB4700772 Sigma

Monoclonal Anti-Neurofilament medium protein antibody produced in mouse

clone NF-09, purified immunoglobulin, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, NE-NI, Primary Antibodies
species reactivity   mammals
application(s)   immunoblotting: suitable
  immunocytochemistry: suitable
  immunohistochemistry: suitable
clone   NF-09, monoclonal
concentration   1 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG2a
shipped in   wet ice
storage temp.   2-8°C
Gene Information   human ... NEFM(4741)
biological source   mouse
conjugate   unconjugated
NCBI accession no.   NM_001105541.1

Description

Immunogen

Pellet of porcine brain cold stable proteins after depolymerization of microtubules.

General description

The antibody NF-09 reacts with both phosphorylated and non-phosphorylated form of medium neurofilament protein (160 kDa) of various species.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?