• USA Home
  • SAB5300465 - Monoclonal Anti-FGG antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB5300465 Sigma

Monoclonal Anti-FGG antibody produced in mouse

clone 5A6, ascites fluid

Synonym: FGG

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, FB-FJ, Primary Antibodies
species reactivity   human
application(s)   direct ELISA: 1:10,000
  indirect immunofluorescence: 1:200-1:1,000
  western blot: 1:500-1:2,000
clone   5A6, monoclonal
antibody form   ascites fluid
isotype   IgG2b
mol wt   mol wt 52 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... FGG(2266)
biological source   mouse
conjugate   unconjugated
NCBI accession no.   NM_000509.4

Description

Immunogen

Purified recombinant fragment of human FGG expressed in E.coli.

Antigen Background

Mouse monoclonal antibody raised against FGG

Physical form

Ascitic fluid containing 0.03% sodium azide.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?