• USA Home
  • SAB5300486 - Monoclonal Anti-EIF4EBP1 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB5300486 Sigma

Monoclonal Anti-EIF4EBP1 antibody produced in mouse

clone 11G12C11, ascites fluid

Synonym: 4E-BP1, 4EBP1, BP-1, MGC4316, PHAS-I

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, E-EI, Primary Antibodies
species reactivity   human
application(s)   direct ELISA: 1:10,000
  immunohistochemistry: 1:200-1:1,000
  western blot: 1:500-1:2,000
clone   11G12C11, monoclonal
antibody form   ascites fluid
isotype   IgG1
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... 4E-BP1(1978)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

Purified recombinant fragment of 4EBP1 expressed in E.coli.

General description

The gene EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1) is mapped to human chromosome 8p12.

Antigen Background

Mouse monoclonal antibody raised against 4E-BP1

Physical form

Ascitic fluid containing 0.03% sodium azide.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Biochem/physiol Actions

The gene EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1) encodes a translation repressor that interacts with eukaryotic translation initiation factor 4E (eIF4E). eIF4E is a multisubunit complex that recruits 40S ribosomal subunits to the 5’ end of mRNA. The interaction of the encoded binding protein with this complex inhibits translation. The phosphorylation of eIF4Ebp1 in response to stimuli such as, UV irradiation and insulin signaling, results in dissociation of this factor from the eIF4E complex, leading to initiation of translation of mRNA. The encoded protein participates in cell proliferation, apoptosis, invasion, and metastasis. It functions as an effector molecule in mTOR (mammalian target of rapamycin complex 1) signaling pathway that regulates protein synthesis. It is found to be overexpressed in hepatocellular carcinoma tissues and serves as a potential prognostic and therapeutic target.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?