• USA Home
  • SAB5300503 - Monoclonal Anti-BRAF antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB5300503 Sigma

Monoclonal Anti-BRAF antibody produced in mouse

clone 1H12, ascites fluid

Synonym: B-RAF1, BRAF1, FLJ95109, RAFB1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, BL-BZ, Primary Antibodies
species reactivity   mouse, human
application(s)   direct ELISA: 1:10,000
  immunohistochemistry: 1:200-1:1,000
  indirect immunofluorescence: 1:200-1:1,000
  western blot: 1:500-1:2,000
clone   1H12, monoclonal
antibody form   ascites fluid
isotype   IgG1
mol wt   mol wt 87 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... BRAF(673)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

Purified recombinant fragment of human BRAF expressed in E.coli.

Antigen Background

Mouse monoclonal antibody raised against BRAF

Physical form

Ascitic fluid containing 0.03% sodium azide.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?