• USA Home
  • SANDTYR - Sandwich Tyrosine Kit

EMAIL THIS PAGE TO A FRIEND
SANDTYR Sigma

Sandwich Tyrosine Kit

Purchase

Properties

shipped in   wet ice
storage temp.   −20°C

Description

Application

Contains tripeptides in which tyrosine is surrounded by various amino acids. Used for UV absorption and fluorescence studies.

For sandwich applications.

Price and Availability

Kit component only

Description

Product #

Add to Cart

Gly-Tyr-Gly SDS    
Leu-Tyr-Leu SDS    
Glu-Tyr-Glu, solid SDS    
Lys-Tyr-Lys SDS    
N-Acetyl-L-Tyrosine Ethyl Ester SDS    
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?