• USA Home
  • SBP3500947 - Rift Valley Fever Virus Polymerase Blocking Peptide for SAB3500947

EMAIL THIS PAGE TO A FRIEND
SBP3500947 Sigma

Rift Valley Fever Virus Polymerase Blocking Peptide for SAB3500947

Synonym: Rift Valley Fever Virus Polymerase Blocking Peptide for SAB3500947

Purchase

Properties

biological source   synthetic
form   liquid
species reactivity   virus
concentration   200 μg/mL
shipped in   wet ice
storage temp.   −20°C

Description

Application

Used as a blocking peptide in immunoblotting applications.

Immunogen

16 amino acids near the amino terminus of Rift Valley Fever Virus Polymerase.

Linkage

This peptide blocks the action of antibody SAB3500947.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

SAB3500947 Anti-RIFT VALLEY FEVER VIRUS POLYMERASE antibody produced in rabbit, affinity isolated antibody

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?