• USA Home
  • SBP3500948 - La Crosse Virus Nucleocapsid Blocking Peptide for SAB3500948

EMAIL THIS PAGE TO A FRIEND
SBP3500948 Sigma

La Crosse Virus Nucleocapsid Blocking Peptide for SAB3500948

Synonym: La Crosse Virus Nucleocapsid Blocking Peptide for SAB3500948

Purchase

Properties

biological source   synthetic
form   liquid
species reactivity   virus
concentration   200 μg/mL
shipped in   wet ice
storage temp.   −20°C

Description

Application

Used as a blocking peptide in immunoblotting applications.

Immunogen

18 amino acids near the center of La Crosse Virus Nucleocapsid.

Linkage

This peptide blocks the action of antibody SAB3500948.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

SAB3500948 Anti-LA CROSSE VIRUS NUCLEOCAPSID antibody produced in rabbit, affinity isolated antibody

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?