• USA Home
  • SBP3500974 - KHDC1 Blocking Peptide for SAB3500974

EMAIL THIS PAGE TO A FRIEND
SBP3500974 Sigma

KHDC1 Blocking Peptide for SAB3500974

Synonym: KHDC1 Blocking Peptide for SAB3500974

Purchase

Properties

biological source   synthetic
form   liquid
species reactivity   human, mouse, rat
concentration   200 μg/mL
shipped in   wet ice
storage temp.   −20°C

Description

Application

Used as a blocking peptide in immunoblotting applications.

Immunogen

15 amino acids near the amino terminus of KHDC1.

Linkage

This peptide blocks the action of antibody SAB3500974.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

SAB3500974 Anti-KHDC1 antibody produced in rabbit, affinity isolated antibody

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?