• USA Home
  • SBP4013 - RPA Interacting Protein (AB1) Blocking Peptide for PRS4013

EMAIL THIS PAGE TO A FRIEND
SBP4013 Sigma

RPA Interacting Protein (AB1) Blocking Peptide for PRS4013

Synonym: RPA Interacting Protein (AB1) Blocking Peptide for PRS4013

Purchase

Properties

biological source   synthetic
form   liquid
species reactivity   human, rat, mouse
concentration   200 μg/mL
shipped in   wet ice
storage temp.   −20°C

Description

Application

Used as a blocking peptide in immunoblotting applications.

Immunogen

17 amino acids near the carboxy terminus of human RPA interacting protein

Linkage

This peptide blocks the action of antibody PRS4013.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

PRS4013 Anti-RPA Interacting Protein (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?