• USA Home
  • SBP4015 - RPA Interacting Protein (AB2) Blocking Peptide for PRS4015

EMAIL THIS PAGE TO A FRIEND
SBP4015 Sigma

RPA Interacting Protein (AB2) Blocking Peptide for PRS4015

Synonym: RPA Interacting Protein (AB2) Blocking Peptide for PRS4015

Purchase

Properties

biological source   synthetic
form   liquid
species reactivity   rat, mouse, human
concentration   200 μg/mL
shipped in   wet ice
storage temp.   −20°C

Description

Application

Used as a blocking peptide in immunoblotting applications.

Immunogen

19 amino acids near the amino terminus of human RPA interacting protein

Linkage

This peptide blocks the action of antibody PRS4015.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

PRS4015 Anti-RPA Interacting Protein (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?