• USA Home
  • SBP4269 - Carabin (AB2) Blocking Peptide for PRS4269

EMAIL THIS PAGE TO A FRIEND
SBP4269 Sigma

Carabin (AB2) Blocking Peptide for PRS4269

Synonym: Carabin (AB2) Blocking Peptide for PRS4269

Purchase

Properties

biological source   synthetic
form   liquid
species reactivity   human, mouse
concentration   200 μg/mL
shipped in   wet ice
storage temp.   −20°C

Description

Application

Used as a blocking peptide in immunoblotting applications.

Immunogen

17 amino acids near the amino terminus of human Carabin.

Linkage

This peptide blocks the action of antibody PRS4269.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

PRS4269 Anti-Carabin (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?