• USA Home
  • SBP4311 - STEAP3 Blocking Peptide for PRS4311

EMAIL THIS PAGE TO A FRIEND
SBP4311 Sigma

STEAP3 Blocking Peptide for PRS4311

Synonym: STEAP3 Blocking Peptide for PRS4311

Purchase

Properties

biological source   synthetic
form   liquid
species reactivity   rat, human, mouse
concentration   200 μg/mL
shipped in   wet ice
storage temp.   −20°C

Description

Application

Used as a blocking peptide in immunoblotting applications.

Immunogen

15 amino acids near the amino terminus of human STEAP3.

Linkage

This peptide blocks the action of antibody PRS4311.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

PRS4311 Anti-STEAP3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?