• USA Home
  • SDS6 - Dalton Mark VI for Weber and Osborn Gels

EMAIL THIS PAGE TO A FRIEND
SDS6 Sigma

Dalton Mark VI for Weber and Osborn Gels

lyophilized powder

Purchase

Properties

Related Categories Molecular Biology, Molecular Weight Markers, Protein Electrophoresis, Proteomics
form   lyophilized powder
storage temp.   2-8°C

Description

Other Notes

Contains a lyophilized mixture of the six proteins plus bromphenol blue tracking dye.

Reconstitution

For Weber and Osborn gels, the marker should be reconstituted with 1 mL phosphate sample buffer [100 mM sodium phosphate, pH 7.0, 1% SDS, 1% (v/v) 2-mercaptoethanol, 6 M urea, and 0.00015% (w/v) bromophenol blue].

Legal Information

Dalton Mark is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


Amicon Pro Purification System
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?