• USA Home
  • SI20100 - MISSION® siRNA Rat Druggable Genome Library

EMAIL THIS PAGE TO A FRIEND
SI20100 Sigma

MISSION® siRNA Rat Druggable Genome Library

6016 targets

Purchase

Properties

Related Categories Functional Genomics and RNAi, MISSION siRNA, MISSION siRNA Rat Druggable Genome, Molecular Biology
storage temp.   −20°C

Description

General description

MISSION siRNA Rat Druggable Genome Library subpanel composed of 6016 therapeutically valuable gene targets designed to support High-Throughput drug screening (HTS) programs. siRNA sequences are optimized for targeting efficiency using the best-in-class design algorithms developed by Rosetta Inpharmatics
MISSION siRNA Library Format:
•  21mer siRNA duplexes with dTdT overhangs
• 3 individual siRNA duplexes per target gene designed using Rosetta Inpharmatics′ design algorithm
• All siRNA duplexes spotted in 96 well microplates with 80 duplexes per plate; delivered dry with a final quantity of 1 nmol per duplex
• Control siRNA sequences included on all plates
• Electronic file supplied including plate maps, target gene list and full sequence information

Legal Information

MISSION is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Kit component also available separately

Description

Product #

Add to Cart

MISSION® siRNA Rat Gene Family Panels, Ligase Panel, 816 targets 1 set SDS SI21100
MISSION® siRNA Rat Gene Family Panels, Kinase Panel, 669 targets 1 set SDS SI22100
MISSION® siRNA Rat Gene Family Panels, Phosphatase Panel, 266 targets 1 set SDS SI23100
MISSION® siRNA Rat Gene Family Panels, Growth Factors/Receptors Panel, 342 targets 1 set SDS SI24100
MISSION® siRNA Rat Gene Family Panels, Cell Adhesion and Cytoskeleton Panel, 433 targets 1 set SDS SI25100
MISSION® siRNA Rat Gene Family Panels, Ion Channel and Transporters Panel, 594 targets 1 set SDS SI26100
MISSION® siRNA Rat Gene Family Panels, Assorted Function Panel, 210 targets 1 set SDS SI27100
MISSION® siRNA Rat Gene Family Panels, GPCR Panel, 275 targets 1 set SDS SI28100
MISSION® siRNA Rat Gene Family Panels, Hydrolase Panel, 188 targets 1 set SDS SI29100
MISSION® siRNA Rat Gene Family Panels, Metabolism and Cell Traffic Panel, 201 targets 1 set SDS SI30100
MISSION® siRNA Rat Gene Family Panels, Nucleic Acid Binding Panel, 287 targets 1 set SDS SI31100
MISSION® siRNA Rat Gene Family Panels, Oxidoreductase Panel, 302 targets 1 set SDS SI32100
MISSION® siRNA Rat Gene Family Panels, Protease Panel, 341 targets 1 set SDS SI33100
MISSION® siRNA Rat Gene Family Panels, Cell Surface and Nuclear Receptors Panel, 647 targets 1 set SDS SI34100
MISSION® siRNA Rat Gene Family Panels, Cell Regulation Panel, 214 targets 1 set SDS SI35100
MISSION® siRNA Rat Gene Family Panels, Transfer and Carrier Proteins Panel, 66 targets 1 set SDS SI36100
MISSION® siRNA Rat Gene Family Panels, Transferase Panel, 174 target 1 set SDS SI37100
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?