• USA Home
  • SI42050 - MISSION® siRNA Mouse Kinase Panel

EMAIL THIS PAGE TO A FRIEND
SI42050 Sigma

MISSION® siRNA Mouse Kinase Panel

623 targets

Purchase

Properties

Related Categories Functional Genomics and RNAi, MISSION siRNA, MISSION siRNA Mouse Kinase Library, Molecular Biology
storage temp.   −20°C

Description

General description

MISSION siRNA Kinase Panel targeting 623 genes of the mouse kinome. MISSION siRNA Library Format:
• 21mer siRNA duplexes with dTdT overhangs
•  3 individual siRNA duplexes per target gene designed using Rosetta Inpharmatics′ design algorithm
•  All siRNA duplexes spotted in 96 well microplates with 80 duplexes per plate; delivered dry with a final quantity of 500 pmol per duplex
•  Control siRNA sequences included on all plates
•  Electronic file supplied including plate maps, target gene list and full sequence information

Legal Information

MISSION is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


Your Key to Success
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?