EMAIL THIS PAGE TO A FRIEND
SIHK0257 Sigma

MISSION® siRNA

Human Kinase CAMK2G (siRNA1), Nano Scale 0.25 nmol

Purchase

Properties

Related Categories AA-CG, Functional Genomics and RNAi, MISSION siRNA, Molecular Biology, siRNA Kinase Nano Scale More...
storage condition   desiccated
NCBI accession no.   NM_172172
storage temp.   −20°C
Gene Information   human ... CAMK2G(818)

Description

General description

The MISSION small scale siRNA are available at 0.25 nmol scale. Sigma′s siRNAs are designed with the Rosetta algorithm and are backed by Sigma-Aldrich’s predesigned siRNA guarantee. You can find all of these off-the-shelf siRNAs on YFG.

Other Notes

Associated siRNA: SASI_Hs01_00152068

Physical form

lyophilized powder

Biochem/physiol Actions

CAMK2G (calcium/calmodulin-dependent protein kinase type II subunit γ), an isoform of CaMKII, is a multifunctional serine/threonine kinase. It mainly controls cell growth and survival in leukemia, lung cancer and liver cancer. In colon cancer cells, it controls apoptosis and autophagy, and is a potential therapeutic target for patients. CAMK2G regulates various receptor/adaptor/kinase/transcription factor interactions by modulating signal pathways involving STAT1 (signal transducer and activator of transcription 1-α/β), NF-κB (nuclear factor), JNK (c-Jun N-terminal kinase), ERK1/2 (extracellular signal-regulated kinase), FOXO1 (forkhead box protein O1), and Wnt/β-catenin.

Legal Information

MISSION is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?