EMAIL THIS PAGE TO A FRIEND
SIHK0275 Sigma

MISSION® siRNA

Human Kinase CARD14 (siRNA3), Nano Scale 0.25 nmol

Purchase

Properties

Related Categories AA-CG, Functional Genomics and RNAi, MISSION siRNA, Molecular Biology, siRNA Kinase Nano Scale More...
storage condition   desiccated
NCBI accession no.   NM_024110
storage temp.   −20°C
Gene Information   human ... CARD14(79092)

Description

General description

The MISSION small scale siRNA are available at 0.25 nmol scale. Sigma′s siRNAs are designed with the Rosetta algorithm and are backed by Sigma-Aldrich’s predesigned siRNA guarantee. You can find all of these off-the-shelf siRNAs on YFG.

Other Notes

Associated siRNA: SASI_Hs01_00165288

Physical form

lyophilized powder

Application

MISSION® siRNA (for CARD14) has been used for silencing of CARD14 (caspase recruitment domain-containing protein 14) in mammalian cells.

Biochem/physiol Actions

CARD14 (caspase recruitment domain-containing protein 14) is a scaffold protein responsible for NF-κB (nuclear factor) signaling in skin keratinocytes. It is mainly needed for maintaining the skin immune homeostasis. Mutations in the gene are associated with pityriasis rubra pilaris (PRP) type V and the development of psoriasis.

Legal Information

M-1000 is a registered trademark of Texaco

MISSION is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?