EMAIL THIS PAGE TO A FRIEND
SIHK1234 Sigma

MISSION® siRNA

Human Kinase MAPKAPK5 (siRNA1), Nano Scale 0.25 nmol

Purchase

Properties

Related Categories FL-MAQ, Functional Genomics and RNAi, MISSION siRNA, Molecular Biology, siRNA Kinase Nano Scale More...
storage condition   desiccated
NCBI accession no.   NM_003668
storage temp.   −20°C
Gene Information   human ... MAPKAPK5(8550)

Description

General description

The MISSION small scale siRNA are available at 0.25 nmol scale. Sigma′s siRNAs are designed with the Rosetta algorithm and are backed by Sigma-Aldrich’s predesigned siRNA guarantee. You can find all of these off-the-shelf siRNAs on YFG.

Other Notes

Associated siRNA: SASI_Hs01_00159255

Physical form

lyophilized powder

Legal Information

MISSION is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?