• USA Home
  • SRP0163 - JMJD2A Active human

EMAIL THIS PAGE TO A FRIEND
SRP0163 Sigma

JMJD2A Active human

recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)

Synonym: JHDM3A, JmjC domain-containing histone demethylation protein 3A, Jumonji domain-containing protein 2A, KIAA0677, Lysine-specific demethylase 4A (KDM4A)

Purchase

Properties

Related Categories Demethylases, Epigenetics, Histone Modifying Enzymes, Molecular Biology, Proteins and Peptides for Epigenetic Research More...
recombinant   expressed in baculovirus infected insect cells
assay   ≥70% (SDS-PAGE)
form   aqueous solution
mol wt   mol wt 56 kDa
NCBI accession no.   NM_014663
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... KDM4A(9682)

Description

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

General description

Human JMJD2A, also known as JHDM3A and KDM4A (GenBank Accession No. NM_014663) (amino acids 1-480) with C-terminal FLAG tag, MW = 56 kDa, expressed in Sf9 cells using a Baculovirus expression system.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Unit Definition

One unit is defined as the amount of enzyme required to demethylate 1 pmol of substrate/min at 37°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 50% glycerol and 3 mM DTT.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Articles

Modulators and Proteins for Methylation

DNA methyltransferases are a family of enzymes that catalyze the transfer of a methyl group to DNA. There are five related DNA cytosine-5-methyltransferases (DNMTs) that transfer a methyl group from ...
Savita Bagga, PhD.
BioFiles v7 n3, 2012, 7–9
Keywords: Gene expression, Methylations, PAGE

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?