• USA Home
  • SRP0180 - Lysyl Oxidase-like 4 (LOXL4) human

EMAIL THIS PAGE TO A FRIEND
SRP0180 Sigma

Lysyl Oxidase-like 4 (LOXL4) human

recombinant, expressed in baculovirus infected insect cells, ≥25% (SDS-PAGE)

Synonym: LOXC, LOXL4, Lysyl oxidase related C

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cytoskeleton and Extracellular Matrix, Extracellular Matrix, Extracellular Matrix Peptides and Proteins More...
recombinant   expressed in baculovirus infected insect cells
assay   ≥25% (SDS-PAGE)
form   aqueous solution
NCBI accession no.   NM_032211
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... LOXL4(84171)

Description

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

General description

Human LOXL4 (GenBank Accession No. NM_032211), amino acids 500-end, MW = 31 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 20% glycerol and 3 mM DTT.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?