• USA Home
  • SRP0214 - PTP1B (1-321)/PTPN1 Active from rat

EMAIL THIS PAGE TO A FRIEND
SRP0214 Sigma

PTP1B (1-321)/PTPN1 Active from rat

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym: PTPN1, Protein tyrosine phosphatase, non-receptor type 1

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Kinase/Phosphatase Biology, Tyrosine Phosphatase (TP), Tyrosine Phosphatase Biology,
recombinant   expressed in E. coli
assay   ≥90% (SDS-PAGE)
form   aqueous solution
mol wt   mol wt 63.3 kDa
NCBI accession no.   NM_012637.2
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... PTPN1(5770)

Description

Application

Useful for the study of enzyme kinetics, regulation, to dephosphorylate target substrates and for screening inhibitors.

General description

Protein tyrosine phosphatase, catalytic domain (amino acids. 1-321), with N-terminal GST tag, MW = 63.3 kDa, expressed in an E. coli expression system.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Unit Definition

One unit will hydrolyze 1 nmol p-nitrophenyl phosphate per minute at pH 7.4 and 30°C.

Physical form

Formulation: recombinant proteins were eluted in 25 mM Tris-HCL, 75 mM NaCl, pH 8.0, 0.05% Tween, 10 mM glutathione, 1 mM DTT and 50% glycerol.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?