EMAIL THIS PAGE TO A FRIEND
SRP0270 Sigma

PDE6C Active human

recombinant, expressed in baculovirus infected insect cells, ≥10% (SDS-PAGE)

Synonym: COD4, Phosphodiesterase 6C, cGMP-specific, cone, alpha prime

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cyclic Nucleotide Metabolism, Cyclic Nucleotide Metabolism Enzymes, G Proteins and Cyclic Nucleotides More...
recombinant   expressed in baculovirus infected insect cells
assay   ≥10% (SDS-PAGE)
form   aqueous solution
mol wt   mol wt 125 kDa
NCBI accession no.   NM_006204
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... PDE6C(5146)

Description

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

General description

Human PDE6C (GenBank Accession No. NM_006204), full length, with N-terminal GST tag, MW = 125 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Unit Definition

One unit is defined as the amount of enzyme that will convert 1 pmole of 3′, 5′-cAMP to 5′ AMP per min at 37°C

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 50% glycerol and 3 mM DTT.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?