ST000037 Sigma

Stemgent® Dox Lentivirus Set: hOKSM conc

verified for composition

DOWNLOAD MSDS (PDF)

Properties

Related Categories Cell Biology, General Stem Cell Biology, Reprogramming Lentiviruses for Human Cells, Stem Cell Biology, Stem Cell Reprogramming More...
impurities   bacteria, tested negative
suitability   verified for composition
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... KLF4(9314), MYC(4609), POU5F1(5460), SOX2(6657)

Description

Frequently Asked Questions

Live Chat and Frequently Asked Questions are available for this Product.

Analysis Note

Viral concentration determined by p24 (capsid antigen) ELISA assay. Transcription factor protein expression verified by immunocytochemistry analysis of transduced human foreskin fibroblasts.

Biochem/physiol Actions

Each doxycycline-inducible lentivirus in this set is capable of expressing one of four human transcription factors (Oct4, Klf4, Sox2 or c-Myc) under the control of the doxycycline (DOX)-inducible tetO operator when transduced into mammalian cells. Each unique virus is provided as a VSV-G pseudotyped and concentrated virus stock capable of infecting both dividing and non-dividing cells. The expression of these four transcription factors, along with the reverse tetracycline transcriptional activator (rtTA), has been shown to reprogram human adult fibroblasts to an embryonic stem (ES) cell-like state known as the induced pluripotent stem (iPS) cell. The GFP-Lentivirus is included as a transduction control.

The Stemgent Dox Lentivirus Set: hOKSM conc was developed in the lab of Rudolf Jaenisch, M.D., at the Whitehead Institute at MIT1. Dr. Jaenisch is a recognized leader in the study of epigenetic regulation of gene expression with numerous publications focused on ES and iPS cellular mechanisms and techniques.

Caution

The greatest safety risk associated with viral delivery systems stems from the potential generation of recombinant viruses that are capable of autonomous replication during the packaging process. This platform eliminates these hazards by combining a disabled viral genome with a unique manufacturing process. Also, gene functions that facilitate the enclosing of the silencing sequence in a viral capsid (e.g., Gag, Pol, Env) are distributed into multiple helper plasmids (which do not contain significant regions of homology) during packaging. This tactic further minimizes the probability of recombination events that might otherwise generate viruses capable of autonomous replication. With these safety measures in place, the Stemgent iPSC Lentivirus particles can be employed in standard Biosafety Level 2 tissue culture facilities and should be treated with the same level of caution as with any other potentially infectious agent. Also, please note that the c-Myc and Klf4 proteins are reported to demonstrate oncogenic properties.

General description

The Stemgent Dox Lentivirus Set: hOKSM provides an affordable inducible method for generating iPS cells. The set includes hOct4, hSox2, hc-Myc, hKlf4, GFP, and the reverse tetracycline transcriptional activator, rtTA, as prepackaged lentivirus particles tested and proven to reprogram human fibroblast cells.

Other Notes

Concentration titer(s) for this product are lot specific. If you have not purchased this product, please contact Sigma′s Technical Service group for the concentration titers. If you have purchased, please consult the online Certificate of Analysis.

Legal Information

For restrictions related to the use of this product, please refer to the accompanying Stemgent Restricted Use License.

Stemgent is a registered trademark of Stemgent, Inc.

Price and Availability

Customers Also Viewed

Doxycycline hyclate

≥98% (TLC)

Stemgent<SUP>®</SUP> Dox Lentivirus Set: hOKSM

verified for composition

Stemgent<SUP>®</SUP> Dox Lentivirus Set: m4F2A

application tested

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?