Popular Documents: Datasheet (PDF)
| Related Categories | Cell Biology, General Stem Cell Biology, Reprogramming Lentiviruses for Human Cells, Stem Cell Biology, Stem Cell Reprogramming More... |
| impurities | bacteria, tested negative |
| suitability | verified for composition |
| shipped in | dry ice |
| storage temp. | −70°C |
| Gene Information | human ... MYC(4609) |
Live Chat and Frequently Asked Questions
are available for this Product.
Viral concentration determined by p24 (capsid antigen) ELISA assay. Transcription factor protein expression verified by immunocytochemistry analysis of transduced human cells.
The expression of the four human transcription factors (TFs) (Oct4, Klf4, Sox2, and c-Myc) has been shown to reprogram human adult fibroblasts to an embryonic stem (ES) cell-like state known as the induced pluripotent stem (iPS) cell. Each unique virus is provided as a VSV-G pseudotyped virus stock capable of efficiently transducing a wide range cell types.
The greatest safety risk associated with viral delivery systems stems from the potential generation of recombinant viruses that are capable of autonomous replication during the packaging process. This platform eliminates these hazards by combining a disabled viral genome with a unique manufacturing process. Also, gene functions that facilitate the enclosing of the silencing sequence in a viral capsid (e.g., Gag, Pol, Env) are distributed into multiple helper plasmids (which do not contain significant regions of homology) during packaging. This tactic further minimizes the probability of recombination events that might otherwise generate viruses capable of autonomous replication. With these safety measures in place, the Stemgent iPSC Lentivirus particles can be employed in standard Biosafety Level 2 tissue culture facilities and should be treated with the same level of caution as with any other potentially infectious agent. Also, please note that the c-Myc and Klf4 proteins are reported to demonstrate oncogenic properties.
The Stemgent Lentivirus hcMyc provides an affordable method for generating iPS cells. The hc-Myc lentivirus, along with the hOct4, hKlf4, and hSox2 prepackaged lentivirus, is tested and proven to reprogram human fibroblast cells.
Concentration titer(s) for this product are lot specific. If you have not purchased this product, please contact Sigma′s Technical Service group for the concentration titers. If you have purchased, please consult the online Certificate of Analysis.
Stemgent is a registered trademark of Stemgent, Inc.
Customers Also Viewed
Production of replication incompetent viral particles
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?