• USA Home
  • T0143 - cis-9-Tetradecenyl acetate

EMAIL THIS PAGE TO A FRIEND
T0143 Sigma

cis-9-Tetradecenyl acetate

≥98%, liquid

Purchase

Properties

assay   ≥98%
form   liquid
storage temp.   −20°C

Description

Packaging

Sealed ampule.

Price and Availability

Safety & Documentation

Safety Information

Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?