• USA Home
  • T0147 - T-2 tetraol tetraacetate

EMAIL THIS PAGE TO A FRIEND
T0147 Sigma

T-2 tetraol tetraacetate

~95%

Synonym: 3α,4β,8α,15-Tetraacetoxy-12,13-epoxytrichothec-9-ene

  • CAS Number 65725-06-6

  • Empirical Formula (Hill Notation) C23H30O10

  • Molecular Weight 466.48

  •  PubChem Substance ID 329826588

Purchase

Properties

InChI Key   GSLBJZJTYRGQOS-RHENDNBISA-N
assay   ~95%
storage temp.   −20°C

Description

Warning

Extremely hazardous! Be aware of the risks and familiar with safety procedures before you use this product.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2811 6.1 / PGI
WGK Germany 
3
RTECS 
YD0107000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?