• USA Home
  • T0154 - Trimethylsilyl-α-D-(+)-glucose

EMAIL THIS PAGE TO A FRIEND
T0154 Sigma

Trimethylsilyl-α-D-(+)-glucose

90-95%, liquid

Synonym: TMS-α-D-(+)-Glucose

  • CAS Number 3327-61-5

  • Empirical Formula (Hill Notation) C21H52O6Si5

  • Molecular Weight 541.06

  •  MDL number MFCD00070519

  •  PubChem Substance ID 24899870

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z, Monosaccharide More...
InChI Key   PPFHNIVPOLWPCF-YMQHIKHWSA-N
assay   90-95%
form   liquid
color   clear colorless
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?