• USA Home
  • T0322 - Nunc® amplification tubes

EMAIL THIS PAGE TO A FRIEND
T0322 Sigma

Nunc® amplification tubes

size 0.2 mL, Strips with matching cap strip

Purchase

Properties

Related Categories Genomics, PCR and Arrays, Molecular Biology, Plant Biotechnology, Plant Laboratory Supplies and Books, Supplies for PCR More...
mfr. no.   Nunc 248151
packaging   case of 600
size   0.2 mL

Description

General description

• Optimized for liquid phase PCR
• GeNunc tubes and caps are made of virgin polypropylene which can withstand temperatures from -20°C to +122°C
• Available in 0.2 mL strips or as 0.2 mL and 0.5 mL individual tubes
• V-shaped tubes with uniformly thin walls
• Dome-shaped lids for good contact with heated lids of cycler
• Compatible with most 0.2 mL and 0.5 thermal cycler formats
• Offers uniform heat transmission for maximum yield
• Certified RNAse and DNAse free

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Protocols

Long and Accurate PCR Amplification of DNA D8045 Protocol

Effective denaturation is accomplished by the use of higher temperatures for shorter periods of time or by the use of co-solvents, such as dimethyl sulfoxide. Addition of DMSO in the reaction at a fi...

Long and Accurate PCR Amplification of DNA with RedAccuTaq D4812 Protocol

Reaction Optimization Reliable amplification of long DNA sequences requires: 1) effective denaturation of DNA template, 2) adequate extension times to produce large products and 3) protection of targ...

Low Contaminant Amplification of DNA Using MTP Taq DNA Polymerase D7442 Protocol

Every precaution should be taken to avoid contamination of reagents with unknown/unwanted DNA. This includes the following:

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?