• USA Home
  • T0324 - Trichloroacetic acid solution

EMAIL THIS PAGE TO A FRIEND
T0324 Sigma

Trichloroacetic acid solution

~6.25 % (w/v), 0.38 N

Synonym: TCA

Purchase

Properties

Related Categories Acids and Bases, Biochemicals and Reagents, Denaturation, Molecular Biology, Protein Sequencing,
InChI Key   YNJBWRMUSHSURL-UHFFFAOYSA-N
concentration   0.38 N
  ~6.25 % (w/v)
storage temp.   2-8°C

Description

Components

Contains 0.025% sodium azide as a preservative

Application

Traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation.
Used as a decalcifier and fixative in microscopy.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2564 8 / PGII
WGK Germany 
2
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?