• USA Home
  • T0447 - Nunc® amplification tubes

EMAIL THIS PAGE TO A FRIEND
T0447 Sigma

Nunc® amplification tubes

size 0.2 mL, with attached caps

Purchase

Description

General description

• Optimized for liquid phase PCR
• GeNunc tubes and caps are made of virgin polypropylene which can withstand temperatures from -20°C to +122°C
• Available in 0.2 mL strips or as 0.2 mL and 0.5 mL individual tubes
• V-shaped tubes with uniformly thin walls
• Dome-shaped lids for good contact with heated lids of cycler
• Compatible with most 0.2 mL and 0.5 thermal cycler formats
• Offers uniform heat transmission for maximum yield
• Certified RNAse and DNAse free

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Optimized PCR-based Detection of Mycoplasma [VIDEO]

The maintenance of contamination-free cell lines is essential to cell-based research. Among the biggest contaminant concerns are mycoplasma contamination. Although mycoplasma do not usually kill cont...
Keywords: Detection methods, Metabolism, Polymerase chain reaction

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?