• USA Home
  • T0646 - Trypsin 1:250 from Porcine Pancreas

EMAIL THIS PAGE TO A FRIEND
T0646 Sigma

Trypsin 1:250 from Porcine Pancreas

1,000-1,500 BAEE units/mg solid

Purchase

Properties

storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
YN5075000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

T4799 Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?