• USA Home
  • T0659 - anti-Tumor necrosis factor-α

EMAIL THIS PAGE TO A FRIEND
T0659 Sigma

anti-Tumor necrosis factor-α

Purchase

Properties

conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?