• USA Home
  • T0690 - Tumor Necrosis Factor Soluble Receptor II (TNF sRII) from mouse

EMAIL THIS PAGE TO A FRIEND
T0690 Sigma

Tumor Necrosis Factor Soluble Receptor II (TNF sRII) from mouse

>97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, cell culture tested

Purchase

Properties

Related Categories Cancer Research, Cell Biology, Cell Signaling and Neuroscience, Cytokines & Growth Factors, Cytokines and Growth Factors,
recombinant   expressed in E. coli
assay   >97% (SDS-PAGE)
form   lyophilized powder
impurities   endotoxin, tested
suitability   cell culture tested
storage temp.   −20°C
Gene Information   mouse ... Tnfrsf1b(21938)

Description

Analysis Note

The biological activity is measured by its ability to inhibit the recombinant mouse TNF-α mediated cytotoxicity in mouse L929 cells, in the presence of the metabolic inhibitor actinomycin D.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile, 0.1% TFA with 2.5 mg bovine serum albumin.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?