• USA Home
  • T0694 - Trizma® Pre-set crystals

EMAIL THIS PAGE TO A FRIEND
T0694 Sigma

Trizma® Pre-set crystals

pH 7.7, average Mw 147.6

Synonym: Tris crystals

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Buffer Convenience Packaging, Buffers A to Z, Good’s Buffers,
mol wt   average Mw 147.6
impurities   DNase, RNase, Protease, NICKase, none detected
  endotoxin, tested
  total aerobic microbial count, tested
  ≤0.05 A290
  ≤0.5% water (Karl Fischer)
  ≤5 ppm Heavy metals (as Pb)
pH   7.40 (37 °C, 0.05 M)(lit.)
  7.70 (25 °C, 0.05 M)(lit.)
  7.7
  8.26 (5 °C, 0.05 M)(lit.)
useful pH range   7.0 - 9.0
suitability   cell culture tested
Featured Industry   Diagnostic Assay Manufacturing

Description

Application

The product is cell culture tested. It is also tested for low endotoxin levels and analyzed for the absence of nucleases and proteases.

Other Notes

Certain pH electrodes do not give accurate results with Tris buffers.

Packaging

In convenient foil pouches

Preparation Note

Trizma solutions can be autoclaved.

Legal Information

Trizma is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?