• USA Home
  • T0889 - Trinitrophenylaminolauroylcholesterol

EMAIL THIS PAGE TO A FRIEND
T0889 Sigma

Trinitrophenylaminolauroylcholesterol

Synonym: 3β-Hydroxy-5-cholestene 3-(12-[([2,4,6-trinitrophenyl)amino]dodecanoate), 5-Cholesten-3β-ol 3-(12-[(2,4,6-trinitrophenyl)amino]dodecanoate), TNPAL-Cholesterol

Purchase

Properties

storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?