• USA Home
  • T0906 - Ultramicro pipette tips
EMAIL THIS PAGE TO A FRIEND
T0906 Sigma
Ultramicro pipette tips
Microcapillary, flat tip, volume range 200 μL, non-sterile
Purchase
Properties
Related Categories Labware, Liquid Handling Equipment, Miscellaneous Pipette Tips, Pipette Tips, Pipettes, Pipettors and Tips More...
material   polypropylene
  colorless tip
sterility   non-sterile
feature   barrier: no
packaging   tray of 200 tips
mfr. no.   Sigma-Aldrich
volume range   200 μL
Description
General description
For loading sample wells in vertical slab gels as thin as 0.4 mm. Mark at 5 μl gives visual confirmation of proper pipet operation. Each tray contains 10 × 20 tips.
Low capacity improves accuracy with small sample volumes by reducing dead space.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?