• USA Home
  • T1008 - Thymidine 3′-monophosphate sodium salt

EMAIL THIS PAGE TO A FRIEND
T1008 Sigma

Thymidine 3′-monophosphate sodium salt

  • CAS Number 108320-91-8

  • Empirical Formula (Hill Notation) C10H14N2O8PNa

  • Molecular Weight 344.19

Purchase

Properties

InChI Key   KTAXSUHEUYNIOC-UHFFFAOYSA-N
storage temp.   −20°C

Description

Preparation Note

Chemically prepared.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?