• USA Home
  • T1159 - 5′-O-Tritylthymidine

EMAIL THIS PAGE TO A FRIEND
T1159 Sigma

5′-O-Tritylthymidine

~98%

  • CAS Number 7791-71-1

  • Empirical Formula (Hill Notation) C29H28N2O5

  • Molecular Weight 484.54

  •  MDL number MFCD00038462

  •  PubChem Substance ID 24899941

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   UYUKMLBIHCJKDF-XJZYHIECSA-N
assay   ~98%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?