• USA Home
  • T1313 - Thayer Martin Medium Base

EMAIL THIS PAGE TO A FRIEND
T1313 Sigma

Thayer Martin Medium Base

pH 7.0±0.2 (25 °C)

Purchase

Properties

Related Categories All Liquid Media/Broths, Media, Microbiology More...
pH   7.0±0.2 (25 °C)

Description

Components

Components (g/L)
Special Peptone 23.00
Starch 1.00
Sodium Chloride 5.00
Agar 13.00

Application

Used as a selective medium for isolating gonococci from pathogenic bacteria.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?