EMAIL THIS PAGE TO A FRIEND
T1406 Sigma

o-Toluidine

liquid

Synonym: 2-Aminotoluene, 2-Methylaniline

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, Natural Dyes, Stains and Dyes
vapor density   3.7 (vs air)
vapor pressure   0.26 mmHg ( 25 °C)
InChI Key   RNVCVTLRINQCPJ-UHFFFAOYSA-N
form   liquid
autoignition temp.   899 °F
color   light yellow
refractive index   n20/D 1.572(lit.)
bp   199-200 °C(lit.)
  89-90 °C/11 mmHg(lit.)
mp   −28 °C(lit.)
solubility   H2O: slightly soluble
density   1.008 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Preparation Note

Processed to minimize the formation of color

Other Notes

This product has been replaced by 185426-ALDRICH | o-Toluidine ≥99%

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1708 6.1 / PGII
WGK Germany 
3
RTECS 
XU2975000
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

185426 o-Toluidine, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?