EMAIL THIS PAGE TO A FRIEND
T1639 Sigma

Trimethyl borate

Synonym: Boric acid trimethyl ester, Methyl borate

Purchase

Properties

density   0.91 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2416 3 / PGII
WGK Germany 
1
RTECS 
ED5600000
Flash Point(F) 
17.6 °F
Flash Point(C) 
-8 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

452920 Trimethyl borate, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?