• USA Home
  • T164 - Testosterone 17β-cypionate

EMAIL THIS PAGE TO A FRIEND
T164 Sigma

Testosterone 17β-cypionate

Synonym: 17β-Hydroxy-4-androsten-3-one 17-cyclopentylpropionate, 4-Androsten-17β-ol-3-one 17-cypionate, Depotestosterone, Depovirin, Testosterone 17β-cyclopentanepropionate

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Biomarkers and Metabolomics Standards, Hormone Standards
InChI Key   HPFVBGJFAYZEBE-ZLQWOROUSA-N
form   neat
drug control   USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
XA3066000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?