• USA Home
  • T1656 - Ultramicro pipette tips
EMAIL THIS PAGE TO A FRIEND
T1656 Sigma
Ultramicro pipette tips
Microcapillary, flat tip, volume range 10 μL, non-sterile
Purchase
Properties
Related Categories Automated Sequencing, Equipment and Accessories, Labware, Liquid Handling Equipment, Miscellaneous Pipette Tips,
material   colorless tip
  polypropylene
sterility   non-sterile
feature   barrier: no
packaging   tray of 10 × 20 tips
mfr. no.   Sigma-Aldrich
volume range   10 μL
Description
Application
For loading sample wells in vertical slab gels as thin as 0.4 mm.
General description
Low capacity improves accuracy with small sample volumes by reducing dead space.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?