• USA Home
  • T1770 - Thiamine monophosphate chloride

EMAIL THIS PAGE TO A FRIEND
T1770 Sigma

Thiamine monophosphate chloride

~98%

Purchase

Properties

assay   ~98%
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

T8637 Thiamine monophosphate chloride dihydrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?