• USA Home
  • T1771 - Troponin from chicken muscle

EMAIL THIS PAGE TO A FRIEND
T1771 Sigma

Troponin from chicken muscle

Lyophilized powder

Purchase

Description

Analysis Note

SDS gel electrophoresis shows the three major bands reported for troponin.

Biochem/physiol Actions

In the absence of Ca2+, inhibits muscle contractility by causing tropomyosin to block the myosin binding sites on actin.

Physical form

Lyophilized powder containing Tris buffer salts

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?