• USA Home
  • T1791 - Hanna heavy-duty thermometer HI 9060
EMAIL THIS PAGE TO A FRIEND
T1791 Sigma
Hanna heavy-duty thermometer HI 9060
Model HI 9060
Purchase
Properties
Related Categories Electronic Meters, Hanna, Hanna Hand-Held Thermometer, Hanna Temperature measurement, Labware,
mfr. no.   Hanna
measuring range   −50-+150 °C temperature, accuracy: 0.3 °C
  −58-+302 °F temperature, accuracy: 0.6 °F
Description
General description
• waterproof design with silicon thermistor sensor
• high/low memory
• temperature hold
• reads °C or °F
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?