• USA Home
  • T1889 - 3′,3′′,5′,5′′-Tetrabromophenolphthalein ethyl ester potassium salt

EMAIL THIS PAGE TO A FRIEND
T1889 Sigma

3′,3′′,5′,5′′-Tetrabromophenolphthalein ethyl ester potassium salt

~90%

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Organic Pollutant Standards, Q-Z, Volatile & Semivolatile Standards More...
InChI Key   WCIQBKUTYDIBJC-UHFFFAOYSA-M
assay   ~90%
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
pH range   3.0 - 5.0, yellow to blue
mp   210 °C (dec.)(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

114774 3′,3′′,5′,5′′-Tetrabromophenolphthalein ethyl ester potassium salt, indicator grade

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?