EMAIL THIS PAGE TO A FRIEND
T1945 Sigma

Trichodion

>95%, solid

  • Empirical Formula (Hill Notation) C12H14O5

  • Molecular Weight 238.24

  •  MDL number MFCD03458796

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, DNA-RNA Transcription Regulators, Gene Regulation and Expression
assay   >95%
form   solid
storage temp.   −20°C

Description

Application

Inhibits NF-κB, αP-1 and STAT1α mediated gene expression induced by IFN-δ resulting in inhibition of COX-2 and NOS ll (iNOS) expression.

Biochem/physiol Actions

Trichodion is a bioactive pyrone that can be isolated from the fungus Trichosporiella sp. Studies have reported that this pyrone can block IFN-γ mediated reporter gene expression in HeLa S3 cells.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?