• USA Home
  • T2024 - Trichloroacetic acid lauryl ester

EMAIL THIS PAGE TO A FRIEND
T2024 Sigma

Trichloroacetic acid lauryl ester

~98%

Synonym: C12

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Organic Pollutant Standards, Q-Z, Volatile & Semivolatile Standards More...
assay   ~98%
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
density   1.12 g/mL at 25 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?